Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl malonimidate dihydrochloride 95%

Supplier: AMBEED, INC
Warning

A549024-100G A549024-10G A549024-1G A549024-250MG A549024-25G A549024-5G
76805-448EA 614 USD InStock
76805-448 76805-450 76805-452 76805-454 76805-456 76805-458
Diethyl malonimidate dihydrochloride 95%
Diethyl malonimidate dihydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₆Cl₂N₂O₂
MW: 231.12 g/mol
MDL Number: MFCD00054633
CAS Number: 10344-69-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty