Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S)-(+)-Camphor-10-sulphonic acid monohydrate 97%

Supplier: AMBEED, INC
Warning

A553535-100G A553535-25G
76805-516EA 44 USD InStock
76805-516 76805-518
(1S)-(+)-Camphor-10-sulphonic acid monohydrate 97%
(1S)-(+)-Camphor-10-sulphonic acid monohydrate
Precaución: for Research Use Only


Formula: C₁₀H₁₆O₄S·1H₂O
MW: 250.32 g/mol
MDL Number: MFCD00150752
CAS Number: 98673-87-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty