Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-(N-Benzyl)-4-iodopyrazole 98%

Supplier: AMBEED, INC
Warning

A557345-100G A557345-100MG A557345-1G A557345-250MG A557345-25G A557345-5G
76805-544EA 420 USD InStock
76805-544 76805-546 76805-548 76805-550 76805-552 76805-554
1H-(N-Benzyl)-4-iodopyrazole 98%
1H-(N-Benzyl)-4-iodopyrazole
Precaución: for Research Use Only


Formula: C₁₀H₉IN₂
MDL Number: MFCD04038745
CAS Number: 50877-42-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty