Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Naphthalene-2-carboxamide 98%

Supplier: AMBEED, INC
Warning

A557429-10G A557429-1G A557429-25G A557429-5G
76805-556EA 49 USD InStock
76805-556 76805-558 76805-560 76805-562
Naphthalene-2-carboxamide 98%
Naphthalene-2-carboxamide
Precaución: for Research Use Only


Formula: C₁₁H₉NO
MDL Number: MFCD00016812
CAS Number: 2243-82-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty