Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Maltitol 95%

Supplier: AMBEED, INC
Warning

A562502-100G A562502-500G A562502-25G
76805-634EA 17 USD InStock
76805-634 76805-636 77307-816
Maltitol 95%
Maltitol
Precaución: for Research Use Only


Formula: C₁₂H₂₄O₁₁
MW: 344.31 g/mol
MDL Number: MFCD00006600
CAS Number: 585-88-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty