Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-aminohexanoate hydrochloride 98%

Supplier: AMBEED, INC
Warning

A562746-1G A562746-25G A562746-5G
76805-638EA 10 USD InStock
76805-638 76805-640 76805-642
Methyl-2-aminohexanoate hydrochloride 98%
Methyl-2-aminohexanoate hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₆ClNO₂
MDL Number: MFCD00058296
CAS Number: 77300-48-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty