Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Methoxycoumarin 97%

Supplier: AMBEED, INC
Warning

A571905-100MG A571905-1G A571905-250MG A571905-25G A571905-5G
76805-718EA 6 USD InStock
76805-718 76805-720 76805-722 76805-724 76805-726 76805-728
7-Methoxycoumarin 97%
7-Methoxycoumarin
Precaución: for Research Use Only


Formula: C₁₀H₈O₃
MDL Number: MFCD00006876
CAS Number: 531-59-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty