Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenylethyl isobutyrate 98%

Supplier: AMBEED, INC
Warning

A579589-1000G A579589-100G A579589-25G A579589-500G
76805-784EA 52 USD InStock
76805-784 76805-786 76805-788 76805-790
2-Phenylethyl isobutyrate 98%
2-Phenylethyl isobutyrate
Precaución: for Research Use Only


Formula: C₁₂H₁₆O₂
MDL Number: MFCD00026441
CAS Number: 103-48-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty