Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dihexyl sulphide 98%

Supplier: AMBEED, INC
Warning

A581738-100G A581738-25G A581738-500G
76805-794EA 26 USD InStock
76805-794 76805-796 76805-798
Dihexyl sulphide 98%
Dihexyl sulphide
Precaución: for Research Use Only


Formula: C₁₂H₂₆S
MW: 202.4 g/mol
Boiling Pt: 230
Melting Pt: -28.3
MDL Number: MFCD00009528
CAS Number: 6294-31-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty