Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Heptadecanoyl chloride 95%

Supplier: AMBEED, INC
Danger

A584267-5G A584267-1G
76805-832EA 68 USD InStock
76805-832 77669-538
Heptadecanoyl chloride 95%
Heptadecanoyl chloride
Precaución: for Research Use Only


Formula: C₁₇H₃₃ClO
MW: 288.90 g/mol
MDL Number: MFCD00000743
CAS Number: 40480-10-2
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty