Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isonicotinoyl chloride hydrochloride 95%

Supplier: AMBEED, INC
Danger

A591052-25G A591052-5G
76805-906EA 19 USD InStock
76805-906 76805-908
Isonicotinoyl chloride hydrochloride 95%
Isonicotinoyl chloride hydrochloride
Precaución: for Research Use Only


Formula: C₆H₄ClNO
MDL Number: MFCD00012830
CAS Number: 39178-35-3
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty