Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Monomethyl maleate 95%

Supplier: AMBEED, INC
Warning

A592700-100G A592700-25G A592700-500G
76805-914EA 54 USD InStock
76805-914 76805-916 76805-918
Monomethyl maleate 95%
Monomethyl maleate
Precaución: for Research Use Only


Formula: C₅H₆O₄
MW: 130.10 g/mol
MDL Number: MFCD00045905
CAS Number: 3052-50-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty