Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetyl-5-bromobenzofuran 95%

Supplier: AMBEED, INC
Warning

A603390-1G A603390-250MG A603390-5G
76805-992EA 30 USD InStock
76805-992 76805-994 76805-996
2-Acetyl-5-bromobenzofuran 95%
2-Acetyl-5-bromobenzofuran
Precaución: for Research Use Only


Formula: C₁₀H₇BrO₂
MDL Number: MFCD00276737
CAS Number: 38220-75-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty