Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Iron(III) bromide 98%

Supplier: AMBEED, INC
Warning

A607565-100G A607565-10G A607565-1G A607565-25G A607565-500G A607565-5G
76806-046EA 103 USD InStock
76806-046 76806-048 76806-050 76806-052 76806-054 76806-056
Iron(III) bromide 98%
Iron(III) bromide
Precaución: for Research Use Only


Formula: Br₃Fe
MW: 295.56 g/mol
MDL Number: MFCD00016082
CAS Number: 10031-26-2
Merck Index: 12,04060
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty