Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Gallamine triethiodide 98%

Supplier: AMBEED, INC
Warning

A616170-100MG A616170-250MG A616170-25MG
76806-100EA 19 USD InStock
76806-100 76806-102 76806-104
Gallamine triethiodide 98%
Gallamine triethiodide
Precaución: for Research Use Only


Formula: C₃₀H₆₀N₃O₃I₃
MDL Number: MFCD00011832
CAS Number: 65-29-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty