Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl Icosanedioate 98%

Supplier: AMBEED, INC
Warning

A643370-1G A643370-25G A643370-5G
76806-326EA 20 USD InStock
76806-326 76806-328 76806-330
Dimethyl Icosanedioate 98%
Dimethyl Icosanedioate
Precaución: for Research Use Only


Formula: C₂₂H₄₂O₄
MW: 370.57 g/mol
MDL Number: MFCD00059302
CAS Number: 42235-38-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty