Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl-5-methoxyisophthalate 98%

Supplier: AMBEED, INC
Warning

A647723-1G A647723-250MG A647723-25G A647723-5G
76806-350EA 49 USD InStock
76806-350 76806-352 76806-354 76806-356
Dimethyl-5-methoxyisophthalate 98%
Dimethyl-5-methoxyisophthalate
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₆
MDL Number: MFCD00272337
CAS Number: 20319-44-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty