Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl vinylphosphonate 97%

Supplier: AMBEED, INC
Danger

A654789-1G A654789-25G A654789-5G A654789-10G
76806-396EA 7 USD InStock
76806-396 76806-398 76806-400 77551-986
Diethyl vinylphosphonate 97%
Diethyl vinylphosphonate
Precaución: for Research Use Only


Formula: C₆H₁₃O₃P
MW: 164.14 g/mol
MDL Number: MFCD00009079
CAS Number: 682-30-4
UN: 3278
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty