Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethylene glycol 99%

Supplier: AMBEED, INC
Warning

A657514-25G A657514-500G A657514-1000G
76806-412EA 5 USD InStock
76806-412 76806-414 77552-002
Ethylene glycol 99%
Ethylene glycol
Precaución: for Research Use Only


Formula: HOCH₂CH₂OH
MDL Number: MFCD00002885
CAS Number: 107-21-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty