Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium methanesulphinate 96%

Supplier: AMBEED, INC
Warning

A657523-1000G A657523-100G A657523-1G A657523-25G A657523-500G A657523-5G
76806-416EA 147 USD InStock
76806-416 76806-418 76806-420 76806-422 76806-424 76806-426
Sodium methanesulphinate 96%
Sodium methanesulphinate
Precaución: for Research Use Only


Formula: CH₃NaO₂S
MW: 102.09 g/mol
MDL Number: MFCD00040392
CAS Number: 20277-69-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty