Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-(2-aminothiazol-4-yl)acetate hydrochloride 98%

Supplier: AMBEED, INC
Warning

A658522-1G A658522-250MG
76806-438EA 20 USD InStock
76806-438 76806-440
Ethyl 2-(2-aminothiazol-4-yl)acetate hydrochloride 98%
Ethyl 2-(2-aminothiazol-4-yl)acetate hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₁ClN₂O₂S
MDL Number: MFCD06795917
CAS Number: 76629-17-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty