Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-methylvalerate 98%

Supplier: AMBEED, INC
Danger

A658624-10G A658624-25G
76806-442EA 15 USD InStock
76806-442 76806-444
Ethyl-2-methylvalerate 98%
Ethyl-2-methylvalerate
Precaución: for Research Use Only


Formula: C₈H₁₆O₂
MW: 144.21 g/mol
MDL Number: MFCD00036564
CAS Number: 39255-32-8
UN: 3272
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty