Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Flavoxate hydrochloride 98%

Supplier: AMBEED, INC
Warning

A666517-1G A666517-1MG A666517-250MG A666517-25G A666517-5G
76806-498EA 32 USD InStock
76806-498 76806-500 76806-502 77669-818 77669-820
Flavoxate hydrochloride 98%
Flavoxate hydrochloride
Precaución: for Research Use Only


Formula: C₂₄H₂₅NO₄
MW: 427.93 g/mol
MDL Number: MFCD00072099
CAS Number: 3717-88-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty