Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isoxazolo[5,4-b]pyridin-3-ylamine 98%

Supplier: AMBEED, INC
Danger

A666882-100MG A666882-1G A666882-250MG A666882-10G
76806-504EA 20 USD InStock
76806-504 76806-506 76806-508 77669-822
Isoxazolo[5,4-b]pyridin-3-ylamine 98%
Isoxazolo[5,4-b]pyridin-3-ylamine
Precaución: for Research Use Only


Formula: C₆H₅N₃O
MDL Number: MFCD04117798
CAS Number: 92914-74-4
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty