Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl (trichloromethyl)phosphonate 97%

Supplier: AMBEED, INC
Warning

A668618-100G A668618-1G A668618-25G A668618-5G
76806-510EA 258 USD InStock
76806-510 76806-512 76806-514 76806-516
Diethyl (trichloromethyl)phosphonate 97%
Diethyl (trichloromethyl)phosphonate
Precaución: for Research Use Only


Formula: C₅H₁₀Cl₃O₃P
MW: 255.46 g/mol
MDL Number: MFCD00013666
CAS Number: 866-23-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty