Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl phenylphosphonite 98% (by GC)

Supplier: AMBEED, INC
Warning

A671386-25G A671386-5G
76806-560EA 39 USD InStock
76806-560 76806-562
Diethyl phenylphosphonite 98% (by GC)
Diethyl phenylphosphonite
Precaución: for Research Use Only


Formula: C₁₀H₁₅O₂P
MW: 198.2 g/mol
MDL Number: MFCD00009086
CAS Number: 1638-86-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty