Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Imidazo[1,2-a]pyridin-3-ylacetic acid 95%

Supplier: AMBEED, INC
Warning

A676888-250MG A676888-100MG
76806-572EA 30 USD InStock
76806-572 77669-852
Imidazo[1,2-a]pyridin-3-ylacetic acid 95%
Imidazo[1,2-a]pyridin-3-ylacetic acid
Precaución: for Research Use Only


Formula: C₉H₈N₂O₂
MW: 176.18 g/mol
MDL Number: MFCD01720477
CAS Number: 17745-04-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty