Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Heptanoic anhydride 95%

Supplier: AMBEED, INC
Danger

A685299-25G A685299-5G
76806-618EA 40 USD InStock
76806-618 77669-860
Heptanoic anhydride 95%
Heptanoic anhydride
Precaución: for Research Use Only


Formula: C₁₄H₂₆O₃
MW: 242.36 g/mol
MDL Number: MFCD00009536
CAS Number: 626-27-7
UN: 1760
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty