Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pseudopelletierine oxime 98%

Supplier: AMBEED, INC
Danger

A694555-100G A694555-25G A694555-5G
76806-660EA 78 USD InStock
76806-660 76806-662 76806-664
Pseudopelletierine oxime 98%
Pseudopelletierine oxime
Precaución: for Research Use Only


Formula: C₉H₁₆N₂O
MDL Number: MFCD03755545
CAS Number: 6164-67-6
UN: 1325
ADR: 4.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty