Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl-2-iodoterephthalate 98%

Supplier: AMBEED, INC
Warning

A706737-1G A706737-250MG A706737-5G
76806-748EA 59 USD InStock
76806-748 76806-750 76806-752
Dimethyl-2-iodoterephthalate 98%
Dimethyl-2-iodoterephthalate
Precaución: for Research Use Only


Formula: C₁₀H₉IO₄
MDL Number: MFCD00060640
CAS Number: 165534-79-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty