Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

H-His-NH₂·2HCl 98%

Supplier: AMBEED, INC
Warning

A719245-1G A719245-25G A719245-5G
76806-824EA 23 USD InStock
76806-824 76806-826 76806-828 76806-830
H-His-NH₂·2HCl 98%
H-His-NH₂·2HCl
Precaución: for Research Use Only


Formula: C₆H₁₂Cl₂N₄O
MDL Number: MFCD00058559
CAS Number: 71666-95-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty