Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl propargylmalonate 97%

Supplier: AMBEED, INC
Warning

A722400-100G A722400-1G A722400-25G A722400-5G
76806-872EA 367 USD InStock
76806-872 76806-874 76806-876 76806-878
Dimethyl propargylmalonate 97%
Dimethyl propargylmalonate
Precaución: for Research Use Only


Formula: C₈H₁₀O₄
MDL Number: MFCD00077856
CAS Number: 95124-07-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty