Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Trimethyl-N-(3-(trifluoromethyl)phenyl)benzenesulphonamide 97%

Supplier: AMBEED, INC
Warning

A732625-10MG A732625-1MG A732625-25MG A732625-50MG A732625-5MG
76807-008EA 46 USD InStock
76807-008 76807-010 76807-012 76807-014 76807-016
2,4,6-Trimethyl-N-(3-(trifluoromethyl)phenyl)benzenesulphonamide 97%
2,4,6-Trimethyl-N-(3-(trifluoromethyl)phenyl)benzenesulphonamide
Precaución: for Research Use Only


Formula: C₁₆H₁₆F₃NO₂S
MW: 343.36 g/mol
MDL Number: MFCD00095824
CAS Number: 200933-14-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty