Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indium(III) iodide 99.999%

Supplier: AMBEED, INC
Danger

A734144-1G A734144-250MG A734144-25G A734144-5G
76807-032EA 18 USD InStock
76807-032 76807-034 76807-036 76807-038
Indium(III) iodide 99.999%
Indium(III) iodide
Precaución: for Research Use Only


Formula: I₃In
MW: 495.53 g/mol
Boiling Pt: 500
Melting Pt: 209.85
MDL Number: MFCD00016151
CAS Number: 13510-35-5
UN: 3288
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty