Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Formanilide 98%

Supplier: AMBEED, INC
Warning

A735961-100G A735961-25G
76807-040EA 40 USD InStock
76807-040 76807-042
Formanilide 98%
Formanilide
Precaución: for Research Use Only


Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 271
MDL Number: MFCD00003276
CAS Number: 103-70-8
Merck Index: 13,04262
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty