Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-((2-(Diethylamino)ethyl)amino)-4-methyl-9H-thioxanthen-9-one 99+%

Supplier: AMBEED, INC
Warning

A737309-100MG A737309-50MG
76807-058EA 285 USD InStock
76807-058 76807-060
1-((2-(Diethylamino)ethyl)amino)-4-methyl-9H-thioxanthen-9-one 99+%
1-((2-(Diethylamino)ethyl)amino)-4-methyl-9H-thioxanthen-9-one
Precaución: for Research Use Only


Formula: C₂₀H₂₄N₂OS
MDL Number: MFCD00866726
CAS Number: 479-50-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty