Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyldiphenylsilane 95%

Supplier: AMBEED, INC
Warning

A739906-100G A739906-1G A739906-25G A739906-5G
76807-090EA 190 USD InStock
76807-090 76807-092 76807-094 76807-096
Methyldiphenylsilane 95%
Methyldiphenylsilane
Precaución: for Research Use Only


Formula: C₁₃H₁₄Si
MW: 198.34 g/mol
Boiling Pt: 267
MDL Number: MFCD00008255
CAS Number: 776-76-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty