Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-bromo-3-hydroxybenzoate 98%

Supplier: AMBEED, INC
Warning

A740850-10G A740850-25G A740850-5G
76807-100EA 67 USD InStock
76807-100 76807-102 76807-104
Ethyl 4-bromo-3-hydroxybenzoate 98%
Ethyl 4-bromo-3-hydroxybenzoate
Precaución: for Research Use Only


Formula: C₉H₉BrO₃
MDL Number: MFCD09038129
CAS Number: 33141-66-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty