Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Manganese phosphate acid P2O5:46-52%

Supplier: AMBEED, INC
Danger

A759177-1000G A759177-100G A759177-500G
76807-216EA 9 USD InStock
76807-216 76807-218 76807-220
Manganese phosphate acid P2O5:46-52%
Manganese phosphate acid
Precaución: for Research Use Only


Formula: H₄MnO₈P₂
MW: 248.91 g/mol
MDL Number: MFCD03453629
CAS Number: 18718-07-5
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty