Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-D-Dab(Aloc)-OH 98%

Supplier: AMBEED, INC
Warning

A774822-100MG A774822-1G A774822-250MG A774822-5G
76807-340EA 10 USD InStock
76807-340 76807-342 76807-344 76807-346
Fmoc-D-Dab(Aloc)-OH 98%
Fmoc-D-Dab(Aloc)-OH
Precaución: for Research Use Only


Formula: C₂₄H₂₇N₂O₆
MW: 424.45 g/mol
MDL Number: MFCD02094487
CAS Number: 387824-78-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty