Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Menthone (mixture of isomers) ≥98% (mixture of isomers)

Supplier: AMBEED, INC
Danger

A775007-25G A775007-1000G
76807-348EA 15 USD InStock
76807-348 76807-350 77670-074
Menthone (mixture of isomers) ≥98% (mixture of isomers)
Menthone (mixture of isomers)
Precaución: for Research Use Only


Formula: C₁₀H₁₈O
MDL Number: MFCD00062998
CAS Number: 10458-14-7
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty