Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl lignocerate 98%

Supplier: AMBEED, INC
Warning

A776628-100MG A776628-1G A776628-250MG
76807-356EA 37 USD InStock
76807-356 76807-358 76807-360
Methyl lignocerate 98%
Methyl lignocerate
Precaución: for Research Use Only


Formula: C₂₅H₅₀O₂
MDL Number: MFCD00009351
CAS Number: 2442-49-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty