Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isradipine ≥98%

Supplier: AMBEED, INC
Warning

A782133-10MG A782133-1MG A782133-25MG A782133-50MG A782133-1G A782133-5G
76807-394EA 6 USD InStock
76807-394 76807-396 76807-398 76807-400 77670-098 77670-100
Isradipine ≥98%
Isradipine
Precaución: for Research Use Only


Formula: C₁₉H₂₁N₃O₅
MW: 371.39 g/mol
MDL Number: MFCD00153820
CAS Number: 75695-93-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty