Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chlormezanone 98%

Supplier: AMBEED, INC
Warning

A792666-1MG A792666-25MG
76807-428EA 6 USD InStock
76807-428 76807-430
Chlormezanone 98%
Chlormezanone
Precaución: for Research Use Only


Formula: C₁₁H₁₂ClNO₃S
MW: 273.73 g/mol
MDL Number: MFCD00143951
CAS Number: 80-77-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty