Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl hexadecanedioate 95%

Supplier: AMBEED, INC
Warning

A796138-1G A796138-5G
76807-460EA 13 USD InStock
76807-460 76807-462
Dimethyl hexadecanedioate 95%
Dimethyl hexadecanedioate
Precaución: for Research Use Only


Formula: C₁₈H₃₄O₄
MW: 314.47 g/mol
MDL Number: MFCD00039806
CAS Number: 19102-90-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty