Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetylbenzofuran 98%

Supplier: AMBEED, INC
Warning

A805059-100G A805059-10G A805059-1G A805059-25G A805059-5G
76807-524EA 163 USD InStock
76807-524 76807-526 76807-528 76807-530 76807-532
2-Acetylbenzofuran 98%
2-Acetylbenzofuran
Precaución: for Research Use Only


Formula: C₁₀H₈O₂
MDL Number: MFCD00005849
CAS Number: 1646-26-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty