Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DL-Isocitric acid-γ-Lactone ≥95%

Supplier: AMBEED, INC
Warning

A809419-100MG A809419-1G A809419-250MG
76807-556EA 23 USD InStock
76807-556 76807-558 76807-560
DL-Isocitric acid-γ-Lactone ≥95%
DL-Isocitric acid-gamma-Lactone
Precaución: for Research Use Only


Formula: C₆H₆O₆
MW: 174.11 g/mol
MDL Number: MFCD00005397
CAS Number: 4702-32-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty