Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Linomide 98%

Supplier: AMBEED, INC
Danger

A809531-100MG A809531-250MG
76807-562EA 173 USD InStock
76807-562 76807-564
Linomide 98%
Linomide
Precaución: for Research Use Only


Formula: C₁₈H₁₆N₂O₃
MDL Number: MFCD00866331
CAS Number: 84088-42-6
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty