Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl violet 97%-103%, Lambert-Beer

Supplier: AMBEED, INC
Danger

A821675-25G A821675-5G
76807-606EA 26.53 USD InStock
76807-606 76807-608 76807-610
Ethyl violet 97%-103%, Lambert-Beer
Ethyl violet
Precaución: for Research Use Only


Formula: C₃₁H₄₂ClN₃
MDL Number: MFCD00011765
CAS Number: 2390-59-2
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty