Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Imidazo[1,2-a]pyrimidine-2-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A846610-100MG A846610-1G A846610-250MG
76807-672EA 17 USD InStock
76807-672 76807-674 76807-676
Imidazo[1,2-a]pyrimidine-2-carboxylic acid 97%
Imidazo[1,2-a]pyrimidine-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₇H₅N₃O₂
MW: 163.14 g/mol
MDL Number: MFCD03419484
CAS Number: 64951-10-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty